Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Disposable Polyethylene Apron
SDP

Catalog No. 19055338
Change view
Click to view available options
Material:
Polyethylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-055-338 Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-055-338 Supplier Keystone Safety Supplier No. APPE4BL2846
Add to Cart
Add to Cart
This item is not returnable. View return policy

Keystone Disposable aprons are made from polymer material that keeps costs down while affording protection from water based liquids.

  • Polyethylene
  • 4 mil.
  • 100% Latex Free
  • Blue
  • 28 x 46 in.
  • These disposable aprons meet FDA guidelines for food handling.
  • Ties in back for a secure fit

Specifications

Type Apron
Size Universal
Material Polyethylene
Color Blue
Disposable Single-use
Closure Type Ties
Sterility Non-sterile
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Dimensions (L x W) 28 x 46 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.