Learn More
GENTECH SOLUTIONS LLC Disposable Reflux Cap, 50mL
Catalog No. NC3728756
Encompass_Preferred
EliteChem Reflux caps are crafted from metals-free polypropylene, making them perfect for digestion process­es requiring the addition of reagents throughout heating. These caps are designed to fit 50-ml digestion tubes and feature a 1/2" (12.7 mm) central hole for convenient reagent addition during the digestion process.
Catalog No. NC3728756 Supplier GENTECH SOLUTIONS LLC Supplier No. 1100200014
EliteChem Reflux caps are crafted from metals-free polypropylene, making them perfect for digestion process­es requiring the addition of reagents throughout heating. These caps are designed to fit 50-ml digestion tubes and feature a 1/2" (12.7 mm) central hole for convenient reagent addition during the digestion process.