Learn More
Andwin Scientific DISPOSABLE RESERVOIR HOLDS UP
SDP
Catalog No. 50819925
Encompass_Preferred
The disposable reservoir features a unique V-shaped design and a wide dimension to fit 12-channel pipettors. This product has a volume capacity of 50 mL and is sold in a case of 100 white polystyrene reservoirs.
Catalog No. 50-819-925 Supplier Andwin Scientific Supplier No. 23202620
The disposable reservoir features a unique V-shaped design and a wide dimension to fit 12-channel pipettors. This product has a volume capacity of 50 mL and is sold in a case of 100 white polystyrene reservoirs.