Learn More
V&P Scientific Inc REAGENT RESERVOIR, Disposable, Polypropylene, 96 Pipettors, V Groove, Sterile, 40/pack
Catalog No. NC2198441
Encompass
REAGENT RESERVIOR, Disposable, Polypropylene, V Groove Designed for 96 Channel Liquid Handling Robots, With Minimal Dead Volume, Sterile, Includes Removable Splash Baffles
This item is not returnable. View return policy
Catalog No. NC2198441 Supplier V&P Scientific Inc Supplier No. VP 792E
REAGENT RESERVIOR, Disposable, Polypropylene, V Groove Designed for 96 Channel Liquid Handling Robots, With Minimal Dead Volume, Sterile, Includes Removable Splash Baffles