Learn More
Braintree Scientific Disposable Safety Scalpel, S/S #12, 10/pkg
SDP
Catalog No. 501954071
Encompass
Disposable Stainless Steel Safety Scalpel - Engineered for safety and simplicity. Complies with the OSHA directive by providing a fully retractable scalpel blade, which protects the user from accidental needle sticks associated with traditional scalpels. Packaged 10 Safety Scalpels per dispenser box, sterile, single use. Reusable sterile stainless dissecting surgical blades and handles also available - call for size and pricing. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4071 Supplier Braintree Scientific Supplier No. 4512
Disposable Stainless Steel Safety Scalpel - Engineered for safety and simplicity. Complies with the OSHA directive by providing a fully retractable scalpel blade, which protects the user from accidental needle sticks associated with traditional scalpels. Packaged 10 Safety Scalpels per dispenser box, sterile, single use. Reusable sterile stainless dissecting surgical blades and handles also available - call for size and pricing. **Sold for research purposes only and not for human use**