Learn More
Sklar Instruments Disposable Scalpel, Curved, Blade, #10, Stainless Steel / Plastic, Disposable, Box of 10.
SDP
Catalog No. 501109708 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Scalpel, Curved, Blade, #10, Stainless Steel / Plastic, Disposable, Box of 10. Sklar's Sterile, Disposable Scalpel #10 is most commonly used for large, straight incisions. It has a curved cutting edge. These disposable scalpels are available in a pack of 10.
Catalog No. 50-110-9708 Supplier Sklar Instruments Supplier No. 063110
Disposable Scalpel, Curved, Blade, #10, Stainless Steel / Plastic, Disposable, Box of 10. Sklar's Sterile, Disposable Scalpel #10 is most commonly used for large, straight incisions. It has a curved cutting edge. These disposable scalpels are available in a pack of 10.