Learn More
Sklar Instruments Disposable Scalpel, Sklar 12, Sklar Sterile, Disposable Scalpel #12, BX10.
SDP
Catalog No. 501094397 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Scalpel, Sklar, #12, Sklar's Sterile, Disposable Scalpel #12 is a small, pointed, crescent shaped blade sharpened along the inside edge of the curve. These disposable scalpels are available in a box of 10.
Catalog No. 50-109-4397 Supplier Sklar Instruments Supplier No. 063112
Disposable Scalpel, Sklar, #12, Sklar's Sterile, Disposable Scalpel #12 is a small, pointed, crescent shaped blade sharpened along the inside edge of the curve. These disposable scalpels are available in a box of 10.