Learn More
Sklar Instruments Disposable Scalpel, Sklar 21, Sklar Sterile, BX10.
SDP
Catalog No. 501094400 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Scalpel, Sklar, #21, Sklar's Sterile, Disposable Scalpel #21 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #21 blade is smaller than the #22 blade and larger than the #20 blade and is compatible with the #4 series and larger handles. This blade is one of the most traditional, commonly used blades and often used for making small incisions in skin and muscle. These disposable scalpels are available in a box of 10.
Catalog No. 50-109-4400 Supplier Sklar Instruments Supplier No. 063121
Disposable Scalpel, Sklar, #21, Sklar's Sterile, Disposable Scalpel #21 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #21 blade is smaller than the #22 blade and larger than the #20 blade and is compatible with the #4 series and larger handles. This blade is one of the most traditional, commonly used blades and often used for making small incisions in skin and muscle. These disposable scalpels are available in a box of 10.