Learn More
Sklar Instruments Disposable Scalpel, Sklar 20, Sklar Sterile, Disposable Scalpel #20 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge.
SDP
Catalog No. 501094399 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Scalpel, Sklar, #20, Sklar's Sterile, Disposable Scalpel #20 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #20 Scalpel Blade is typically not used in surgery, but may be if a long incision or digit amputation is necessary.
Catalog No. 50-109-4399 Supplier Sklar Instruments Supplier No. 063120
Disposable Scalpel, Sklar, #20, Sklar's Sterile, Disposable Scalpel #20 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #20 Scalpel Blade is typically not used in surgery, but may be if a long incision or digit amputation is necessary.