Learn More
Sklar Instruments Disposable Scalpel, Sklar 22. Sklar Sterile, BX10.
SDP
Catalog No. 501094401 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Scalpel, Sklar, #22. Sklar's Sterile, Disposable Scalpel #22 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #22 blade is larger than both the #20 and #21 blades. It is compatible with the #4 series and larger handles. This blade is commonly used in cardiac and thoracic procedures to make incisions in the bronchus.These disposable scalpels are available in a box of 10.
Catalog No. 50-109-4401 Supplier Sklar Instruments Supplier No. 063122
Disposable Scalpel, Sklar, #22. Sklar's Sterile, Disposable Scalpel #22 is a larger version on the #10 Scalpel Blade. It has a curved cutting edge and a flat, unsharpened back edge. The #22 blade is larger than both the #20 and #21 blades. It is compatible with the #4 series and larger handles. This blade is commonly used in cardiac and thoracic procedures to make incisions in the bronchus.These disposable scalpels are available in a box of 10.