Learn More
Sklar Instruments Disposable Scalpel, Sklar 23. Sklar Sterile, Box of 10.
SDP
Catalog No. 501094402 Shop All Sklar Instruments Products
Encompass_Preferred
Disposable Scalpel, Sklar, #23. Sklar's Sterile, Disposable Scalpel #23 is a leaf-shaped blade sharpened along the leading edge. It is commonly used for making long incisions. These disposable scalpels are available in a box of 10.
Catalog No. 50-109-4402 Supplier Sklar Instruments Supplier No. 063123
Disposable Scalpel, Sklar, #23. Sklar's Sterile, Disposable Scalpel #23 is a leaf-shaped blade sharpened along the leading edge. It is commonly used for making long incisions. These disposable scalpels are available in a box of 10.