Learn More
Electron Microscopy Sciences Disposable Scalpel, Mini, #15
SDP
Catalog No. 50949468
Encompass_Preferred
Feather Disposable Mini Scalpels are made from heat-treated stainless steel. The blades are smaller than standard blades, come with chemical resistant, slender plastic handle. Handle is 5 3/4" (145 mm) long. ISO 14001 compliant, ISO 9001 certified and ISO13485 certified.
Catalog No. 50-949-468 Supplier Electron Microscopy Sciences Supplier No. 7204115
Feather Disposable Mini Scalpels are made from heat-treated stainless steel. The blades are smaller than standard blades, come with chemical resistant, slender plastic handle. Handle is 5 3/4" (145 mm) long. ISO 14001 compliant, ISO 9001 certified and ISO13485 certified.