Learn More
World Precision Instrument Disposable Scalpel, #15, Sterile, 10/Box
SDP
Catalog No. 50822460
Encompass
The surgical stainless steel blades are installed on plastic handles, and each unit comes sterile and individually wrapped. These are perfect for one time surgical uses, dissections, crafts, student use, woodworking, taxidermy, clay modeling and more.
Each disposable knife comes with a cap to protect the blade for safe disposal or to save the used scalpel for another application.
The plastic handle of the scalpel is textured to ensure a firm grip. It is lightweight for comfortable use. And the handle includes a handy, integrated 6 cm metric ruler.
Catalog No. 50-822-460 Supplier World Precision Instrument Supplier No. 500351
The surgical stainless steel blades are installed on plastic handles, and each unit comes sterile and individually wrapped. These are perfect for one time surgical uses, dissections, crafts, student use, woodworking, taxidermy, clay modeling and more.
Each disposable knife comes with a cap to protect the blade for safe disposal or to save the used scalpel for another application.
The plastic handle of the scalpel is textured to ensure a firm grip. It is lightweight for comfortable use. And the handle includes a handy, integrated 6 cm metric ruler.