Learn More
Research Products International Corp Disposable Scalpel, Size 10, Sterile, Individually Wrapped, 10 per Case
SDP
Catalog No. 50998780
Encompass
Scalpels are used for a variety of tasks that require precise cutting and dissection. Size 10: Features a large, curved blade with a rounded cutting edge, commonly used for making long, smooth incisions in soft tissue. It is often used in general  dissection.
Common uses include:
Tissue Dissection: Scalpels are used to carefully dissect tissues from organs or other biological samples in cell culture, histology, and molecular biology.
Sample Preparation: In molecular biology and genetics, scalpels are used to excise sections of tissue or samples for DNA/RNA extraction or protein analysis.
Microdissection: For isolating specific cells or small structures under a microscope in fields like neuroscience, oncology, or developmental biology.
Catalog No. 50-998-780 Supplier Research Products International Corp Supplier No. 248180
Scalpels are used for a variety of tasks that require precise cutting and dissection. Size 10: Features a large, curved blade with a rounded cutting edge, commonly used for making long, smooth incisions in soft tissue. It is often used in general  dissection.
Common uses include:
Tissue Dissection: Scalpels are used to carefully dissect tissues from organs or other biological samples in cell culture, histology, and molecular biology.
Sample Preparation: In molecular biology and genetics, scalpels are used to excise sections of tissue or samples for DNA/RNA extraction or protein analysis.
Microdissection: For isolating specific cells or small structures under a microscope in fields like neuroscience, oncology, or developmental biology.