Learn More
Exel International Disposable Scalpels
Catalog No. 1484002
Change view
Click to view available options
Size:
# 10
# 11
# 15
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
14-840-02 # 15
14-840-00 # 10
14-840-01 # 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-840-02 Supplier Exel International Supplier No. 29556
Add to Cart
Add to Cart
This item is not returnable. View return policy
Item is Sterile and individually wrapped
Specifications
Blade Material Stainless Steel
Product Type Scalpel
Size # 15
Disposable Single-use
Sterility Sterile
Certifications/Compliance ISO 2859, ISO7740, BS 2982