Learn More
Andwin Scientific DISPOSABLE SCALPELS 10PK
SDP
Catalog No. NC0608802
Encompass_Preferred
The Feather Disposable Scalpels are attached with a blade and a handle. Each scalpel is individually wrapped in aluminum wrapper to ensure its cleanliness. This product is also disposable in order to eliminate the chance of contamination. The blade is in style #10.
Catalog No. NC0608802 Supplier Andwin Scientific Supplier No. 82029850
The Feather Disposable Scalpels are attached with a blade and a handle. Each scalpel is individually wrapped in aluminum wrapper to ensure its cleanliness. This product is also disposable in order to eliminate the chance of contamination. The blade is in style #10.