Learn More
BURKLE INC Scoop LaboPlast, single-use, PS, 5.07 oz. (150 ml), with cover
Catalog No. 503081369
Encompass_Preferred
The LaboPlast disposable scoop, made from premium white polystyrene, is produced in ISO Class 7 cleanroom conditions, individually packaged to ensure maximum cleanliness.
Designed for sensitive sampling of powders, granulates, and pastes, this ergonomic scoop features a slightly raised front edge for easy, spill-free use across various materials.
Its disposable design eliminates the need for cleaning or sterilization, saving time and resources while enhancing lab efficiency and staff safety by reducing exposure to hazardous substances.
Individually sealed packaging keeps each scoop contamination-free until use, preventing cross-contamination and ensuring sample integrity by single-use disposal.
Catalog No. 50-308-1369 Supplier BURKLE INC Supplier No. 53780008
The LaboPlast disposable scoop, made from premium white polystyrene, is produced in ISO Class 7 cleanroom conditions, individually packaged to ensure maximum cleanliness.
Designed for sensitive sampling of powders, granulates, and pastes, this ergonomic scoop features a slightly raised front edge for easy, spill-free use across various materials.
Its disposable design eliminates the need for cleaning or sterilization, saving time and resources while enhancing lab efficiency and staff safety by reducing exposure to hazardous substances.
Individually sealed packaging keeps each scoop contamination-free until use, preventing cross-contamination and ensuring sample integrity by single-use disposal.