Learn More
BURKLE INC Sampling scoop, long handle, 1.69 oz. (50 ml), PS, white
Catalog No. 503081193
Encompass_Preferred
The disposable sample scoop with extra-long handle is designed for single use, ideal for sampling from large containers, even with limited contents or narrow openings.
The straight edge allows precise sampling directly from the container bottom, ensuring accuracy and ease.
Perfect for powders, granules, and pastes, this scoop suits various materials and applications.
Its disposable design saves time by eliminating cleaning or sterilization, boosting lab and production efficiency. It also enhances safety by reducing exposure to hazardous substances and avoiding handling of contaminated reusable tools.
Individually packaged to stay contamination-free until use, it prevents cross-contamination between samples. After use, simply dispose to maintain process integrity.
Catalog No. 50-308-1193 Supplier BURKLE INC Supplier No. 53785013
The disposable sample scoop with extra-long handle is designed for single use, ideal for sampling from large containers, even with limited contents or narrow openings.
The straight edge allows precise sampling directly from the container bottom, ensuring accuracy and ease.
Perfect for powders, granules, and pastes, this scoop suits various materials and applications.
Its disposable design saves time by eliminating cleaning or sterilization, boosting lab and production efficiency. It also enhances safety by reducing exposure to hazardous substances and avoiding handling of contaminated reusable tools.
Individually packaged to stay contamination-free until use, it prevents cross-contamination between samples. After use, simply dispose to maintain process integrity.