Learn More
Lakeland Industries Disposable Shirt
Supplier:  Lakeland Industries CTL2013X
Encompass_Preferred
MicroMax NS provides the protection you need from dirt, grease, grime, light chemical splash and blood. Long sleeve design with a snap front. Available Sizes: MD, 4X, 5X
Catalog No. 50-284-0495