Learn More
Lakeland Industries Disposable Sleeves
Catalog No. 502841114
Encompass_Preferred
Lakeland's MicroMax NS fabric offers breathable, reliable protection against dirt, grease, grime, and light chemical splash. The CTL850P-18 disposable sleeves are offered in 18" length.
Catalog No. 50-284-1114 Supplier Lakeland Industries Supplier No. CTL850P18
Lakeland's MicroMax NS fabric offers breathable, reliable protection against dirt, grease, grime, and light chemical splash. The CTL850P-18 disposable sleeves are offered in 18" length.