Learn More
Electron Microscopy Sciences Disposable Slide Staining Tray Set
SDP
Catalog No. 501926622
Encompass_Preferred
Stain, rinse, and dry your slides on a single working tray. Use the tray for a clean and fresh working surface area for staining. These trays are made from Polypropylene / polyethylene blend and is very durable.
The deep well can hold liquid waste up to 38mL. Place eight slides comfortably onto the base. With its compact size, tray provides comfortable transport from work area to sink. Convenient pour spouts to easily dispose of unwanted liquid waste. Unique dark lid protects slides when used with fluorescents. The tray holds up to 8 slides maximum with a 38ml liquid capacity. A complete set has 4 base trays and one lid. The dark lid protects the slides for light sensitive applications.
Catalog No. 50-192-6622 Supplier Electron Microscopy Sciences Supplier No. 71147
Stain, rinse, and dry your slides on a single working tray. Use the tray for a clean and fresh working surface area for staining. These trays are made from Polypropylene / polyethylene blend and is very durable.
The deep well can hold liquid waste up to 38mL. Place eight slides comfortably onto the base. With its compact size, tray provides comfortable transport from work area to sink. Convenient pour spouts to easily dispose of unwanted liquid waste. Unique dark lid protects slides when used with fluorescents. The tray holds up to 8 slides maximum with a 38ml liquid capacity. A complete set has 4 base trays and one lid. The dark lid protects the slides for light sensitive applications.