Learn More
Sigma Aldrich Fine Chemicals Biosciences smartSpatula
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5600497500EA
Encompass
The smartSpatula is a clean economical and disposable spatula for chemical and biological material transfer collection and manipulation. Available with double ended design has spoon/spatula on one end and a pick to break up clumps at the other or as a micro design with both ends as pick design.The Macro smartSpatula is 50% larger and has a long shaft to ease access to material in larger containers. The flexible spatula ends are shatter resistant after liquid nitrogen freezing and readily form to an object to scrape or collect samples. Made of polypropylene. Autoclavable.
The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc
Catalog No. 50-291-4691