Learn More
Fisherbrand™ Disposable SMS Laboratory Coats
Catalog No. 14901010 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
White
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
14-901-010 Blue X-Large
14-901-000 White Small
14-901-001 White Medium
14-901-002 White Large
14-901-003 White X-Large
14-901-004 White 2X-Large
14-901-005 White 3X-Large
14-901-006 White 5X-Large
14-901-007 Blue Small
14-901-008 Blue Medium
14-901-009 Blue Large
14-901-011 Blue 2X-Large
14-901-012 Blue 3X-Large
14-901-013 Blue 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 14-901-010
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Fisherbrand™ Disposable SMS Lab Coats come complete with a knitted collar and cuffs. Designed to protect healthcare professionals, lab technicians and other users from fluids and cross-contamination.
Fisherbrand™ premium protective apparel is available in both lab coats and hip length jackets. Made from high quality, ultra-soft SMS fabric designed to keep you protected and comfortable.
  • Fluid-resistant material meets AAMI L2 standards
  • 42 GSM high quality, breathable SMS material
  • Anti-static coating
  • High quality front snap button closure
  • Comfortable knit collar and cuffs
  • 3-pocket design
  • Convenient side slits for easy access to pockets
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Liquids
Style Knee-length
No. of Pockets 3
Collar Type Knit
Latex Free Yes
Certifications/Compliance ANSI, AAMI PB70
Packaging Type Bagged
Autoclavable Not Autoclavable
No. per Pack 10/Pk.