Learn More
Fisherbrand™ Disposable SMS Laboratory Jackets
Catalog No. 14901019 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
White
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
14-901-019 White 3X-Large
14-901-014 White Small
14-901-015 White Medium
14-901-016 White Large
14-901-017 White X-Large
14-901-018 White 2X-Large
14-901-020 White 5X-Large
14-901-021 Blue Small
14-901-022 Blue Medium
14-901-023 Blue Large
14-901-024 Blue X-Large
14-901-025 Blue 2X-Large
14-901-026 Blue 3X-Large
14-901-027 Blue 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Catalog No. 14-901-019
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Fisherbrand™ Disposable SMS Lab Jackets with a knitted collar and cuffs. Designed to protect healthcare professionals, lab technicians and other users from fluids and cross-contamination.
Fisherbrand™ premium protective apparel is available in both lab coats and hip length jackets. Made from high quality, ultra-soft SMS fabric designed to keep you protected and comfortable.
  • Fluid-resistant material meets AAMI L2 standards
  • 42 GSM high quality, breathable SMS material
  • Anti-static coating
  • High quality front snap button closure
  • Comfortable knit collar and cuffs
  • 3-pocket design
  • Convenient hip length
Specifications
Product Type General Purpose Lab Coat
Size 3X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Liquids
Style Hip-length
No. of Pockets 3
Collar Type Knit
Latex Free Yes
Certifications/Compliance ANSI, AAMI PB70
Packaging Type Bagged
Autoclavable Not Autoclavable
No. per Pack 10/Pk.