Learn More
METTLER TOLEDO 3P INDUSTRIAL DISPOSABLE SOFTENING CUP 50-PK
Catalog No. NC1678952
Encompass
  • Disposable softening cups for efficient softening point determination.
  • Reduces cleaning time associated with reusable containers.
  • Single-use aluminum construction enhances workflow.
  • Diameter of 6.35 mm for compatibility with standard equipment.
  • Pack contains 50 cups for high-volume use.
  • No cleaning requirement after use.
  • Economic option for laboratory applications.
  • Minimizes exposure to harmful solvents.
Catalog No. NC1678952 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 30027448
  • Disposable softening cups for efficient softening point determination.
  • Reduces cleaning time associated with reusable containers.
  • Single-use aluminum construction enhances workflow.
  • Diameter of 6.35 mm for compatibility with standard equipment.
  • Pack contains 50 cups for high-volume use.
  • No cleaning requirement after use.
  • Economic option for laboratory applications.
  • Minimizes exposure to harmful solvents.