Learn More
Cole-Parmer Disposable Stir Bar, PTFE-coated alnico V magnet, 1/2" x 1/8", 100/pk
Catalog No. NC9627640 Shop All Cole-Parmer Products
Encompass_Preferred
These stir bars are ideal for demanding chemical and biological applications where cross-contamination is a concern. They have strong alnico V magnets with a one-piece coating of PTFE and priced to be disposable. The stir bars are offered in a convenient bulk package of 100 stir bars.
Catalog No. NC9627640 Supplier Cole-Parmer Supplier No. 0476902
These stir bars are ideal for demanding chemical and biological applications where cross-contamination is a concern. They have strong alnico V magnets with a one-piece coating of PTFE and priced to be disposable. The stir bars are offered in a convenient bulk package of 100 stir bars.