Learn More
Surgical Design™ Disposable Surgical Blade Remover
Catalog No. 21124675 Shop All Surgical Design Products
Change view
Click to view available options
Capacity (English):
100 Blades
Catalog No. Capacity (English)
21-124-675 100 Blades
1 options
Catalog No. 21-124-675 Supplier Surgical Design™ Supplier No. BRDP
Add to Cart
Add to Cart
Disposable surgical blade remover enables hands-free scalpel blade removal to help minimize sharps injuries in surgical, pathology, and postmortem settings.
  • Plastic blade removal container for safe disposal of scalpel blades
  • Enables hands-free blade removal, reducing risk of sharps injury
  • Compatible with a wide range of standard scalpel handles (3, 4, and specialty patterns)
  • Designed for surgical, laboratory, pathology, and autopsy environments
  • Internal locking mechanism secures the blade once detached
  • Lightweight, compact, and easy to use with gloved hands
  • Clear visual confirmation of blade capture
  • Fully disposable to maintain sterile workflow and minimize contamination
  • Durable medical-grade plastic construction
  • Helps comply with sharps handling safety protocols
Specifications
Capacity (English) 100 Blades
Color Green
Disposable Limited-use
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Plastic
Product Type Sharps Disposal Box