Learn More
Exel International Disposable Surgical Blades
Description
• Made from premium quality Swiss Carbon Steel and British Stainless Steel
• Computer-controlled grinding, sharpening and polishing ensures consistently smooth incision properties
• Sterilized by Gamma radiation 2.5m-RADS
• A Vapor Corrosion Inhibitor (VCI) paper lining, leakage testing, and an aluminum foil peel pack help ensure quality and integrity when ready for use
• Each production lot is sampled and conforms to ISO 2859, inspection level (S4) AQL 1.0 and 2.5
• Blade rigidity is tested to eliminate blade breakage
• Blades conform to ISO 7740 (1995) and BS 2982 (1986)
• Blades are ultrasonically cleaned prior to packing
Specifications
Specifications
| Blade Material | Stainless Steel |
| Product Type | Surgical Blade |
| Size | # 15 |
| Disposable | Single-use |
| Sterility | Sterile |