Learn More
Exel International Disposable Surgical Blades
Catalog No. 1484024
Change view
Click to view available options
Size:
# 10
# 11
# 15
# 22
Blade Material:
Carbon Steel
Stainless Steel
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Blade Material
14-840-24 # 15 Carbon Steel
14-840-15 # 10 Stainless Steel
14-840-22 # 11 Carbon Steel
14-840-18 # 15 Stainless Steel
14-840-16 # 11 Stainless Steel
14-840-20 # 22 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 14-840-24 Supplier Exel International Supplier No. 29532
Add to Cart
Add to Cart
This item is not returnable. View return policy
•    Made from premium quality Swiss Carbon Steel and British Stainless Steel
•    Computer-controlled grinding, sharpening and polishing ensures consistently smooth incision properties
•    Sterilized by Gamma radiation 2.5m-RADS
•    A Vapor Corrosion Inhibitor (VCI) paper lining, leakage testing, and an aluminum foil peel pack help ensure quality and integrity when ready for use
•    Each production lot is sampled and conforms to ISO 2859, inspection level (S4) AQL 1.0 and 2.5
•    Blade rigidity is tested to eliminate blade breakage
•    Blades conform to ISO 7740 (1995) and BS 2982 (1986)
•    Blades are ultrasonically cleaned prior to packing
Specifications
Blade Material Carbon Steel
Product Type Surgical Blade
Size # 15
Disposable Single-use
Sterility Sterile