Learn More
Research Products International Corp Disposable Tissue Grinder with Vitrified Tip, Fits 15ml Tubes, 10 per Case
SDP
Catalog No. 501977500
Encompass
Closed system tissue grinders are design to provide maximum safety. Tissues are ground inside of a sealed container thereby reducing risk of sample contact or exposure to aerosols. Narrow pestle eliminates the possibility of sample overflow. Pestle styles include a standard molded abrasive surface or vitrified tip for more aggressive tissue grinding. Tissue grinders are fully assembled with pestle and sample tube. Supplied in easy to open sterile packs.
Catalog No. 50-197-7500 Supplier Research Products International Corp Supplier No. 141914
Closed system tissue grinders are design to provide maximum safety. Tissues are ground inside of a sealed container thereby reducing risk of sample contact or exposure to aerosols. Narrow pestle eliminates the possibility of sample overflow. Pestle styles include a standard molded abrasive surface or vitrified tip for more aggressive tissue grinding. Tissue grinders are fully assembled with pestle and sample tube. Supplied in easy to open sterile packs.