Learn More
Kent Scientific Disposable Towel Drapes for SurgiSuite Multi-Functional Surgical Platforms
SDP
Catalog No. 13005198 Shop All Kent Scientific Products
Encompass
Change view
Click to view available options
Dimensions (L x W):
45.7 x 66 cm (18 x 26 in.)
Catalog No. Dimensions (L x W)
13-005-198 45.7 x 66 cm (18 x 26 in.)
1 options
Catalog No. 13-005-198 Supplier Kent Scientific Supplier No. SURGI50233
For use with SurgiSuite and SurgiSuite-LG Multi-Functional Surgical Platforms.
  • Made of poly-interlined, three-ply construction (tissue/poly/tissue)
  • Provides absorbency while preventing possible moisture or bacteria migration
  • Sterile
Specifications
For Use With (Application) Surgery, Homeothermic Warming, Surgical Procedures, Tissue Retraction
Type Towel Drapes
Disposable Single-use
For Use With (Equipment) SurgiSuite and SurgiSuite-LG Multi-Functional Surgical Platforms
Sterility Sterile
Dimensions (L x W) 45.7 x 66 cm (18 x 26 in.)