Learn More
Andwin Scientific DISPOSABLE TRANSFER PIPETS
SDP
Catalog No. NC0461460
Encompass_Preferred
The disposable 7.5 mL transfer pipet is made from see-through low density polyethylene that is long-lasting as it will not shatter or break. This product is classified as nontoxic and inert to most acids. It is also compatible to use for liquid oxygen. This transfer pipet features a uniform drop size and a low-affinity surface that minimizes the loss of cells and important proteins as a result of binding.
Catalog No. NC0461460 Supplier Andwin Scientific Supplier No. 16001188
The disposable 7.5 mL transfer pipet is made from see-through low density polyethylene that is long-lasting as it will not shatter or break. This product is classified as nontoxic and inert to most acids. It is also compatible to use for liquid oxygen. This transfer pipet features a uniform drop size and a low-affinity surface that minimizes the loss of cells and important proteins as a result of binding.