Learn More
Alkali Scientific ASI™ Disposable Tweezers, PS, Sterile, 123mm, Individual Wrapped,50/Pk, Sharp
SDP
Catalog No. 503027369
Encompass_Preferred
ASI™ Disposable Tweezers are made of clean and durable polystyrene material, and it is four times lighter than conventional metal tweezers, reducing the strain on the wrist during the experiment. It also minimizes cross-contamination of the sample. The sharp type is suitable for handling fine samples, and the broad type is optimized for handling cover glass or plastic coverslip.
Catalog No. 50-302-7369 Supplier Alkali Scientific Supplier No. DTS123
ASI™ Disposable Tweezers are made of clean and durable polystyrene material, and it is four times lighter than conventional metal tweezers, reducing the strain on the wrist during the experiment. It also minimizes cross-contamination of the sample. The sharp type is suitable for handling fine samples, and the broad type is optimized for handling cover glass or plastic coverslip.