Learn More
Azzota Corp Disposable Uv-Vis Fluorometer Cuvette - 3.5Ml X 100 Square Cuvettes 10mm X 10mm X 45mm Four Sides Clear Methacrylate Pathlength: 10mm 3.5Ml Volume Wavelength Range: 285-800Nm (Uv-Vis)