Learn More
Azzota Corp Disposable Vis Fluorometer Cuvette - 3.5Ml X 100 Square Cuvettes 10mm X 10mm X 45mm Four Sides Clear Polystyrene Pathlength: 10mm 3.5Ml Volume Wavelength Range: 340-800Nm (Vis) Package Of 100
Catalog No. 507532899
Encompass_Preferred
Disposable Vis Fluorometer Cuvette - 3.5ml x 100 Square Cuvettes 10mm x 10mm x 45mm Four sides clear Polystyrene Pathlength: 10mm 3.5ml volume Wavelength Range: 340-800nm (Vis) Package of 100
Catalog No. 50-753-2899 Supplier Azzota Corp Supplier No. A912
Disposable Vis Fluorometer Cuvette - 3.5ml x 100 Square Cuvettes 10mm x 10mm x 45mm Four sides clear Polystyrene Pathlength: 10mm 3.5ml volume Wavelength Range: 340-800nm (Vis) Package of 100