Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Disposal Bag w/Biohazard symbol and print

Catalog No. 07000604 Shop All Greiner Bio-One Products
Change view
Click to view available options
Dimensions (L x W):
780 x 600mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
07-000-604 780 x 600mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-604 Supplier Greiner Bio-One Supplier No. 646203
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For hygienic disposal of contaminated laboratory items. Greiner Bio-One Disposal Bag is available with w/Biohazard symbol and print on the bag.

  • Non-sterile
  • Polypropylene (PP) suitable for steam autoclaves
  • Nominal capacities from 10L to 130L
  • For single use only
TRUSTED_SUSTAINABILITY

Specifications

Description w/Biohazard symbol and print
Autoclavable Autoclavable
Closure Type None
Color Translucent
Disposable Single-use
For Use With (Application) Hygienic disposal of contaminated laboratory items
Material Polypropylene
Sterility Non-sterile
Dimensions (L x W) 780 x 600mm
Length (English) 30.7 in.
Width (English) 23.6 in.
Product Type Biohazard Autoclave Bag
Capacity (Metric) 65 L
Length (Metric) 780 mm
Width (Metric) 600 mm
Thickness (Metric) 0.05 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.