Learn More
Rheosense Inc Viscometer pipette, 400 µL, 100/pack
SDP
Catalog No. NC1559672
Encompass
Pipette is designed for use with the microVISC portable, small sample viscometer, 400 µL volume.
Catalog No. NC1559672 Supplier Rheosense Inc Supplier No. H1002
Pipette is designed for use with the microVISC portable, small sample viscometer, 400 µL volume.