Learn More
Grainger Bausch & Lomb Lens Cleaning Wipes, Pre-Moistened, Individually Wrapped, 5 in x 8 in, 100/Pk.
Catalog No. NC9784054
Encompass_Preferred
These pre-moistened lens cleaning tissues are designed to effectively remove dust, dirt, and smudges from a wide range of surfaces, including eyeglasses, safety goggles, face shields, and other personal eyewear. Formulated to provide anti-static and anti-fog properties, these tissues ensure clear vision on both glass and plastic lenses.
  • Pre-moistened for convenient, single-step cleaning
  • Individually wrapped for portability and ease of use
  • Anti-static and anti-fog formula prevents haze and streaks
  • Silicone-free, butoxyethanol-free, ethanol-free, and methanol-free
  • Compatible with both glass and plastic lenses, including camera lenses and monitor screens
  • Packaged in a dispenser box suitable for wall mounting or countertop placement
  • Dimensions: 5 in. x 8 in.
This item is not returnable. View return policy
Catalog No. NC9784054 Supplier Grainger Supplier No. 2AR69
These pre-moistened lens cleaning tissues are designed to effectively remove dust, dirt, and smudges from a wide range of surfaces, including eyeglasses, safety goggles, face shields, and other personal eyewear. Formulated to provide anti-static and anti-fog properties, these tissues ensure clear vision on both glass and plastic lenses.
  • Pre-moistened for convenient, single-step cleaning
  • Individually wrapped for portability and ease of use
  • Anti-static and anti-fog formula prevents haze and streaks
  • Silicone-free, butoxyethanol-free, ethanol-free, and methanol-free
  • Compatible with both glass and plastic lenses, including camera lenses and monitor screens
  • Packaged in a dispenser box suitable for wall mounting or countertop placement
  • Dimensions: 5 in. x 8 in.