Learn More
Waters Corp DisQuE Tube, 150 mg MgSO4, 50 mg PSA, 2 mL
Catalog No. 502392450 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clean-up tube is used in QuEChERS methods for multi-pesticide residue analysis in food. It offers a simple and straightforward sample preparation technique for various food and agricultural products, enabling direct analysis by GC-MS or LC-MS/MS technology.
  • Contains 150 mg magnesium sulfate and 50 mg primary secondary amine bonded silica.
  • Used in the clean-up step of QuEChERS methods to remove matrix interferences from extracts.
  • Suitable for multi-residue analysis of pesticides, veterinary drugs, and mycotoxins.
  • Optimized for fruits and vegetables with high water content; additional water can be added for low water content products.
  • Saves time and effort in sample preparation.
  • Conveniently packaged in a foil pouch and centrifuge tube.
Catalog No. 50-239-2450 Supplier Waters Corp Supplier No. 186004572
This clean-up tube is used in QuEChERS methods for multi-pesticide residue analysis in food. It offers a simple and straightforward sample preparation technique for various food and agricultural products, enabling direct analysis by GC-MS or LC-MS/MS technology.
  • Contains 150 mg magnesium sulfate and 50 mg primary secondary amine bonded silica.
  • Used in the clean-up step of QuEChERS methods to remove matrix interferences from extracts.
  • Suitable for multi-residue analysis of pesticides, veterinary drugs, and mycotoxins.
  • Optimized for fruits and vegetables with high water content; additional water can be added for low water content products.
  • Saves time and effort in sample preparation.
  • Conveniently packaged in a foil pouch and centrifuge tube.