Learn More
Waters Corp Waters Sample Cleanup Tube, 900 mg MgSO4 & 150 mg PSA, 15 mL, 50 pk
Catalog No. 50791211 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 15 mL clean-up tube is designed for larger extract volume clean-up in QuEChERS methods, specifically for multi-pesticide residue analyses in food using the QUECHERS CEN Method 15662. It enables direct analysis via GC-MS or LC-MS/MS with simple dilution, saving time and effort.
  • Used for multi-pesticide residue analyses in food.
  • Designed for larger extract volume clean-up.
  • Allows direct analysis via GC-MS or LC-MS/MS technology with simple dilution.
  • Provides significant time and effort savings.
  • Quick and straightforward sample preparation approach.
  • Suitable for multi-residue analysis of pesticides, veterinary medicines, and mycotoxins in food and agricultural goods.
  • Packaged in a handy foil packet and includes a centrifuge tube.
  • Suitable for high water content fruits and vegetables; adding water improves extraction for low water content goods.
Catalog No. 50-791-211 Supplier Waters Corp Supplier No. 186004833
This 15 mL clean-up tube is designed for larger extract volume clean-up in QuEChERS methods, specifically for multi-pesticide residue analyses in food using the QUECHERS CEN Method 15662. It enables direct analysis via GC-MS or LC-MS/MS with simple dilution, saving time and effort.
  • Used for multi-pesticide residue analyses in food.
  • Designed for larger extract volume clean-up.
  • Allows direct analysis via GC-MS or LC-MS/MS technology with simple dilution.
  • Provides significant time and effort savings.
  • Quick and straightforward sample preparation approach.
  • Suitable for multi-residue analysis of pesticides, veterinary medicines, and mycotoxins in food and agricultural goods.
  • Packaged in a handy foil packet and includes a centrifuge tube.
  • Suitable for high water content fruits and vegetables; adding water improves extraction for low water content goods.