Learn More
Waters Corp Waters DisQuE Tube, MgSO4/PSA/C18, 2 mL, 100 pk
Catalog No. 50791208 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clean-up tube streamlines sample preparation for multi-residue analysis, effective for extracts from QUECHERS AOAC Method 2007.01. It uses C18 for high-fat food samples and supports multi-pesticide residue analyses in food, veterinary medicines, and mycotoxins.
  • Used for extracts from QUECHERS AOAC Method 2007.01, step 1.
  • d-SPE tube with C18 for high-fat food samples.
  • Suitable for multi-pesticide, veterinary medicine, and mycotoxin analyses.
  • Offers quick and straightforward sample preparation.
  • Includes foil bag and centrifuge tube.
  • Enables direct analysis via GC-MS or LC-MS/MS with simple dilution.
  • Incorporates enhanced extraction technology for time savings.
  • Optimized for high water content produce; adaptable for low water content goods by adding water.
Catalog No. 50-791-208 Supplier Waters Corp Supplier No. 186004830
This clean-up tube streamlines sample preparation for multi-residue analysis, effective for extracts from QUECHERS AOAC Method 2007.01. It uses C18 for high-fat food samples and supports multi-pesticide residue analyses in food, veterinary medicines, and mycotoxins.
  • Used for extracts from QUECHERS AOAC Method 2007.01, step 1.
  • d-SPE tube with C18 for high-fat food samples.
  • Suitable for multi-pesticide, veterinary medicine, and mycotoxin analyses.
  • Offers quick and straightforward sample preparation.
  • Includes foil bag and centrifuge tube.
  • Enables direct analysis via GC-MS or LC-MS/MS with simple dilution.
  • Incorporates enhanced extraction technology for time savings.
  • Optimized for high water content produce; adaptable for low water content goods by adding water.