Learn More
Waters Corp DisQuE Tube, 900 mg MgSO4, 150 mg PSA, C18
Supplier: Waters Corp 186004834
This clean-up tube is engineered for multi-pesticide residue analyses in food, designed for larger extract volume clean-up in QuEChERS CEN Method 15662. It incorporates C18 for high-fat food samples and is supplied as a 50-pack of 15 mL tubes with d-SPE components.
- Clean-up tube for QuEChERS CEN Method 15662, Step 1 extract
- Packaged in a foil pouch and centrifuge tube
- Allows direct analysis of final extract by GC-MS or LC-MS/MS technology with simple dilution
- Offers considerable savings of time and effort
- Extraction can be optimized for products with low water content by adding more water
- Suitable for multi-residue analysis for pesticides, veterinary drugs, and mycotoxins