Learn More
Waters Corp Waters DisQuE Sample Preparation Kit, CEN Method 15662, QuEChERS, 100 pk
Catalog No. 50791068 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This dispersive sample preparation kit is designed for the QuEChERS Method CEN Method 15662, enabling multi-pesticide residue analyses in food. It provides a comprehensive solution for simplified dispersive solid-phase extraction (d-SPE). The kit contains 100 units of both extraction and clean-up tubes with pre-weighed sorbents and buffers, adhering to regulatory methods.
  • Designed for QuEChERS Method CEN Method 15662.
  • Includes both extraction and clean-up tubes.
  • Each 50 mL extraction tube contains 4 g MgSO4, 1 g NaCl, 1 g trisodium citrate dehydrate, and 0.5 g disodium hydrogencitrate sesquihydrate.
  • Each 2 mL clean-up tube contains 150 mg MgSO4, 25 mg PSA (primary-secondary amine), and 25 mg C18.
  • Comes with detailed method instructions.
Catalog No. 50-791-068 Supplier Waters Corp Supplier No. 176001903
This dispersive sample preparation kit is designed for the QuEChERS Method CEN Method 15662, enabling multi-pesticide residue analyses in food. It provides a comprehensive solution for simplified dispersive solid-phase extraction (d-SPE). The kit contains 100 units of both extraction and clean-up tubes with pre-weighed sorbents and buffers, adhering to regulatory methods.
  • Designed for QuEChERS Method CEN Method 15662.
  • Includes both extraction and clean-up tubes.
  • Each 50 mL extraction tube contains 4 g MgSO4, 1 g NaCl, 1 g trisodium citrate dehydrate, and 0.5 g disodium hydrogencitrate sesquihydrate.
  • Each 2 mL clean-up tube contains 150 mg MgSO4, 25 mg PSA (primary-secondary amine), and 25 mg C18.
  • Comes with detailed method instructions.