Learn More
Waters Corp Waters DisQuE Pouch, AOAC Method, 50 mL
Catalog No. 50466501 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The DisQuE AOAC Method 1.5 g Sodium Acetate and 6 g MgSO4, 50 mL Pouch provides essential salts for the QuEChERS AOAC Method 2007.01 extraction step, utilizing acetic acid as a buffer. It offers efficient sample preparation for multi-residue analysis of pesticides, veterinary drugs, and mycotoxins across food and agricultural products. Each pack contains 50 pouches, meticulously packaged in a foil pouch to prevent contamination.
  • Designed for simple and straightforward sample preparation.
  • Addresses deficiencies of traditional extraction methods for pesticides in fruits and vegetables.
  • Facilitates direct analysis of the final extract by GC-MS or LC-MS/MS technology with simple dilution.
  • Saves time and effort in the analytical process.
  • Optimized for multi-residue pesticide analysis of fruits and vegetables with high water content.
Catalog No. 50-466-501 Supplier Waters Corp Supplier No. 186006812
The DisQuE AOAC Method 1.5 g Sodium Acetate and 6 g MgSO4, 50 mL Pouch provides essential salts for the QuEChERS AOAC Method 2007.01 extraction step, utilizing acetic acid as a buffer. It offers efficient sample preparation for multi-residue analysis of pesticides, veterinary drugs, and mycotoxins across food and agricultural products. Each pack contains 50 pouches, meticulously packaged in a foil pouch to prevent contamination.
  • Designed for simple and straightforward sample preparation.
  • Addresses deficiencies of traditional extraction methods for pesticides in fruits and vegetables.
  • Facilitates direct analysis of the final extract by GC-MS or LC-MS/MS technology with simple dilution.
  • Saves time and effort in the analytical process.
  • Optimized for multi-residue pesticide analysis of fruits and vegetables with high water content.