Learn More
Waters Corp Waters DisQuE Reagent, QuEChERS Method, 50 mL Pouch
Catalog No. 50466502 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This QuEChERS reagent pouch contains salts for the extraction step in CEN Method 15662, ideal for multi-residue analysis of pesticides, veterinary drugs, and mycotoxins in various food and agricultural products. Its sorbent pouch allows direct extract analysis by GC-MS or LC-MS/MS with simple dilution, significantly cutting sample preparation time. The extraction can be optimized for low water content products by adding supplemental water.
  • Contains salts for QuEChERS CEN Method 15662 extraction.
  • Designed for multi-residue analysis of pesticides, veterinary drugs, and mycotoxins in food and agricultural products.
  • Sorbent pouch enables direct extract analysis by GC-MS or LC-MS/MS with simple dilution.
  • Conveniently packaged in a foil pouch, ensuring product integrity.
  • Saves time and effort in sample preparation.
  • Optimizable for low water content products by adding additional water.
Catalog No. 50-466-502 Supplier Waters Corp Supplier No. 186006813
This QuEChERS reagent pouch contains salts for the extraction step in CEN Method 15662, ideal for multi-residue analysis of pesticides, veterinary drugs, and mycotoxins in various food and agricultural products. Its sorbent pouch allows direct extract analysis by GC-MS or LC-MS/MS with simple dilution, significantly cutting sample preparation time. The extraction can be optimized for low water content products by adding supplemental water.
  • Contains salts for QuEChERS CEN Method 15662 extraction.
  • Designed for multi-residue analysis of pesticides, veterinary drugs, and mycotoxins in food and agricultural products.
  • Sorbent pouch enables direct extract analysis by GC-MS or LC-MS/MS with simple dilution.
  • Conveniently packaged in a foil pouch, ensuring product integrity.
  • Saves time and effort in sample preparation.
  • Optimizable for low water content products by adding additional water.