Learn More
Waters Corp Waters Corporation Sample Preparation Kit, QuEChERS Method, AOAC 2007.01, Pouch Format
Catalog No. 50465947 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for Dispersive Sample Preparation using the QuEChERS Method AOAC 2007.01. It provides 100 units, each containing a salt pouch, an empty 50 mL centrifuge tube, and a clean-up tube. Salt pouches contain 6 g magnesium sulfate and 1.5 g sodium acetate, while 2 mL clean-up tubes include 150 mg magnesium sulfate and 50 mg PSA.
  • Designed for dSPE using QuEChERS Method AOAC 2007.01.
  • Includes 100 units: salt pouches, empty 50 mL centrifuge tubes, and clean-up tubes.
  • Salt pouches contain 6 g magnesium sulfate and 1.5 g sodium acetate.
  • Clean-up tubes contain 150 mg magnesium sulfate and 50 mg PSA.
  • Streamlines environmental sample preparation for LC analysis.
  • No specialized equipment needed; accessible for all expertise levels.
  • Compatible with various LC methods.
  • Employs dSPE to remove interferences and concentrate analytes.
Catalog No. 50-465-947 Supplier Waters Corp Supplier No. 176002922
This kit is designed for Dispersive Sample Preparation using the QuEChERS Method AOAC 2007.01. It provides 100 units, each containing a salt pouch, an empty 50 mL centrifuge tube, and a clean-up tube. Salt pouches contain 6 g magnesium sulfate and 1.5 g sodium acetate, while 2 mL clean-up tubes include 150 mg magnesium sulfate and 50 mg PSA.
  • Designed for dSPE using QuEChERS Method AOAC 2007.01.
  • Includes 100 units: salt pouches, empty 50 mL centrifuge tubes, and clean-up tubes.
  • Salt pouches contain 6 g magnesium sulfate and 1.5 g sodium acetate.
  • Clean-up tubes contain 150 mg magnesium sulfate and 50 mg PSA.
  • Streamlines environmental sample preparation for LC analysis.
  • No specialized equipment needed; accessible for all expertise levels.
  • Compatible with various LC methods.
  • Employs dSPE to remove interferences and concentrate analytes.