Learn More
Waters Corp Waters Sample Preparation Kit, CEN Method, Pouch Format, 100 Kits
Catalog No. 50465948 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This dispersive sample preparation kit is designed for the QuEChERS method CEN Method 15662. It includes 100 units each of salt pouches, empty 50 mL centrifuge tubes, and clean-up tubes for efficient multi-residue pesticide analysis in food samples.
  • Each pouch contains 1 g trisodium citrate dihydrate, 0.5 g disodium hydrogencitrate sesquihydrate, 1 g NaCl, and 4 g MgSO4.
  • Empty 50 mL centrifuge tube included in each kit.
  • Each 2 mL clean-up tube contains 150 mg MgSO4, 25 mg PSA, and 25 mg C18.
  • Supports CEN Method 15662 for QuEChERS extraction.
  • Optimized for multi-residue pesticide analysis in fruits and vegetables.
  • Facilitates extraction from commodities with low water content by allowing additional water.
  • Aids in the clean-up of pigmented samples by allowing the addition of Graphitized Carbon Black (GCB).
Catalog No. 50-465-948 Supplier Waters Corp Supplier No. 176002923
This dispersive sample preparation kit is designed for the QuEChERS method CEN Method 15662. It includes 100 units each of salt pouches, empty 50 mL centrifuge tubes, and clean-up tubes for efficient multi-residue pesticide analysis in food samples.
  • Each pouch contains 1 g trisodium citrate dihydrate, 0.5 g disodium hydrogencitrate sesquihydrate, 1 g NaCl, and 4 g MgSO4.
  • Empty 50 mL centrifuge tube included in each kit.
  • Each 2 mL clean-up tube contains 150 mg MgSO4, 25 mg PSA, and 25 mg C18.
  • Supports CEN Method 15662 for QuEChERS extraction.
  • Optimized for multi-residue pesticide analysis in fruits and vegetables.
  • Facilitates extraction from commodities with low water content by allowing additional water.
  • Aids in the clean-up of pigmented samples by allowing the addition of Graphitized Carbon Black (GCB).