Learn More
Research Products International Corp Disruption Beads for Yeast/Fungi, 0.5mm Bead Size, Zirconia/Silica Beads, 3.7 g/cc Density, 1 Pound Bottle
SDP
Catalog No. 50212146
Encompass
Disruption Beads for homogenizing, grinding and disintegrating biological samples using a BeadBeater and screw cap micro-tube.
Beads are unwashed and should be washed prior to using.
Catalog No. 50-212-146 Supplier Research Products International Corp Supplier No. 9834
Disruption Beads for homogenizing, grinding and disintegrating biological samples using a BeadBeater and screw cap micro-tube.
Beads are unwashed and should be washed prior to using.