Learn More
Research Products International Corp Disruption Beads for Bacteria, 0.1mm Bead Size, Glass Beads, 2.5 g/cc Density, 1 Pound Bottle
SDP
Catalog No. 50212142
Encompass
Disruption Beads for homogenizing, grinding and disintegrating biological samples using a BeadBeater and screw cap micro-tube.
Beads are unwashed and should be washed prior to using.
Catalog No. 50-212-142 Supplier Research Products International Corp Supplier No. 9830
Disruption Beads for homogenizing, grinding and disintegrating biological samples using a BeadBeater and screw cap micro-tube.
Beads are unwashed and should be washed prior to using.