Learn More
Research Products International Corp Disruption Beads for Fibrous Plant Tissue, 2.3mm Bead Size, Chrome Steel Beads, 7.9 g/cc Density
SDP
Catalog No. 501367618
Encompass
Disruption Beads for homogenizing, grinding and disintegrating biological samples using a BeadBeater and screw cap micro-tube.
Beads are unwashed and should be washed prior to using.
Catalog No. 50-136-7618 Supplier Research Products International Corp Supplier No. 9839
Disruption Beads for homogenizing, grinding and disintegrating biological samples using a BeadBeater and screw cap micro-tube.
Beads are unwashed and should be washed prior to using.